(3-(Diethylamino)propyl)trimethoxysilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N-Diethyl-3-(trimethoxysilyl)propylamin, Trimethoxy(3-(diethylamino)propyl)silan, (3-Diethylaminopropyl)trimethoxysilan

M-Clarity™ Qualitätsstufe = MQ100

448664-5ML
SIAL448664-5MLEA 88.4 EUR
SIAL448664-5ML
(3-(Diethylamino)propyl)trimethoxysilan, Sigma-Aldrich®
(3-(Diethylamino)propyl)trimethoxysilan
Formel: (H₃CCH₂)₂NCH₂CH₂CH₂Si(OCH₃)₃
Molecular Weight: 235,4 g/mol
MDL: MFCD00039882
CAS-Nummer: 41051-80-3
EINECS: 255-192-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR