Anisol-D8 (98% D), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

448818-1G
SIAL448818-1GEA 411 EUR
SIAL448818-1G
Anisol-D8 (98% D), Sigma-Aldrich®
Anisol-D8
Formel: C₇D₈O
MDL: MFCD00084119
CAS-Nummer: 54887-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR