1H,1H,2H,2H-Perfluoroctyltrichlorsilan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
448931-10G
SIAL448931-10GEA
123
EUR
SIAL448931-10G
1H,1H,2H,2H-Perfluoroctyltrichlorsilan, Sigma-Aldrich®
1H,1H,2H,2H-Perfluoroctyltrichlorsilan
Formel:
C₈H₄Cl₃F₁₃Si |
MDL:
MFCD00042363 CAS-Nummer: 78560-45-9 EINECS: 278-947-6 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...