Methyl-2-naphthoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-naphthoate

M-Clarity™ Qualitätsstufe = MQ200

449075-5G
SIAL449075-5GEA 83.9 EUR
SIAL449075-5G
Methyl-2-naphthoat, Sigma-Aldrich®
Methyl-2-naphthoat
Formel: C₁₂H₁₀O₂
Molecular Weight: 186,21 g/mol
MDL: MFCD00017345
CAS-Nummer: 2459-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR