Dikaliumhexachloroiridat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Kaliumhexachloroiridat(IV)
M-Clarity™ Qualitätsstufe = MQ100
450162-1G
SIAL450162-1GEA
233
EUR
SIAL450162-1G
Dikaliumhexachloroiridat, Sigma-Aldrich®
Dikaliumhexachloroiridat
Formel:
Cl₆IrK₂ Molecular Weight: 483,13 g/mol |
MDL:
MFCD00011369 CAS-Nummer: 16920-56-2 EINECS: 240-976-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...