Aluminiumsulfat Hydrat, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Aluminum(III) sulphate xhydrate

M-Clarity™ Qualitätsstufe = MQ100

450308-5G
SIAL450308-5GEA 78.7 EUR
SIAL450308-5G
Aluminiumsulfat Hydrat, Sigma-Aldrich®
Aluminiumsulfat Hydrat
Formel: Al₂(SO₄)₃·xH₂O
Molecular Weight: 366,21 g/mol
MDL: MFCD00149136
CAS-Nummer: 17927-65-0
EINECS: 233-135-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR