N-Boc-anilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

450359-25G
SIAL450359-25GEA 105 EUR
SIAL450359-25G
N-Boc-anilin, Sigma-Aldrich®
N-Boc-anilin
Formel: C₁₁H₁₅NO₂
MDL: MFCD00043723
CAS-Nummer: 3422-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR