Kupfer(II)sulfid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Kupfersulfid
M-Clarity™ Qualitätsstufe = MQ100
450820-10G
SIAL450820-10GEA
212
EUR
SIAL450820-10G
Kupfer(II)sulfid, Sigma-Aldrich®
Kupfer(II)sulfid
Formel:
CuS Molecular Weight: 95,61 g/mol Dichte: 4,6 g/cm³ (20 °C) |
MDL:
MFCD00016066 CAS-Nummer: 1317-40-4 EINECS: 215-271-2 Merck Index: 12,02724 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...