Scandium(III)chlorid Hexahydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

451274-1G
SIAL451274-1GEA 92.2 EUR
SIAL451274-1G
Scandium(III)chlorid Hexahydrat, Sigma-Aldrich®
Scandium(III)chlorid Hexahydrat
Formel: Cl₃Sc·6H₂O
Molecular Weight: 259,41 g/mol
MDL: MFCD00149852
CAS-Nummer: 20662-14-0
EINECS: 233-799-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR