Benzo[a]pyren-[D12] (98% D), Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Benzo[a]pyrene-[²H₁₂]

M-Clarity™ Qualitätsstufe = MQ200

451797-10MG
SIAL451797-10MGEA 483 EUR
SIAL451797-10MG
Benzo[a]pyren-[D12] (98% D), Sigma-Aldrich®
Benzo[a]pyren-[D12]
Formel: C₂₀D₁₂
CAS-Nummer: 63466-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR