3,5-Dibrom-4-hydroxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-Carbaldehyd-2,6-dibromphenol

M-Clarity™ Qualitätsstufe = MQ100

453137-5G
SIAL453137-5GEA 59.9 EUR
SIAL453137-5G
3,5-Dibrom-4-hydroxybenzaldehyd, Sigma-Aldrich®
3,5-Dibrom-4-hydroxybenzaldehyd
Formel: C₇H₄Br₂O₂
Molecular Weight: 279,92 g/mol
MDL: MFCD00016980
CAS-Nummer: 2973-77-5
EINECS: 221-017-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR