3-Bromphenetol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-3-ethoxybenzol, (3-Bromphenyl)ethylether

453250-5ML
SIAL453250-5MLEA 85.1 EUR
SIAL453250-5ML
3-Bromphenetol, Sigma-Aldrich®
3-Bromphenetol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00274287
CAS-Nummer: 2655-84-7
EINECS: 220-186-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR