4-Cyanphenylhydrazin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydrazinylbenzonitrile hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

453471-5G
SIAL453471-5GEA 108 EUR
SIAL453471-5G
4-Cyanphenylhydrazin Hydrochlorid, Sigma-Aldrich®
4-Cyanphenylhydrazin Hydrochlorid
Formel: C₇H₈ClN₃
Molecular Weight: 169,61 g/mol
MDL: MFCD00673994
CAS-Nummer: 2863-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR