2-Brom-5-nitroanilin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
454257-5G
SIAL454257-5GEA
85.6
EUR
SIAL454257-5G
2-Brom-5-nitroanilin, Sigma-Aldrich®
2-Brom-5-nitroanilin
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217,02 g/mol Schmelzpunkt: 141 °C |
MDL:
MFCD00051578 CAS-Nummer: 10403-47-1 EINECS: 233-874-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...