2-Fluor-5-nitrobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

454281-1G
SIAL454281-1GEA 35 EUR
SIAL454281-1G
2-Fluor-5-nitrobenzaldehyd, Sigma-Aldrich®
2-Fluoro-5-nitrobenzaldehyde
Formel: C₇H₄FNO₃
Molecular Weight: 169,11 g/mol
MDL: MFCD00042298
CAS-Nummer: 27996-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR