2,2'-Thenil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Di(thiophen-2-yl)ethane-1,2-dione

M-Clarity™ Qualitätsstufe = MQ100

454389-5G
SIAL454389-5GEA 160 EUR
SIAL454389-5G
2,2'-Thenil, Sigma-Aldrich®
2,2'-Thenil
Formel: C₁₀H₆O₂S₂
Molecular Weight: 222,29 g/mol
MDL: MFCD00173678
CAS-Nummer: 7333-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR