(R)-(-)-Glycidyl-methylether, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

454680-1G 454680-5G
SIAL454680-1GEA 91.8 EUR
SIAL454680-1G SIAL454680-5G
(R)-(-)-Glycidyl-methylether, Sigma-Aldrich®
(R)-(-)-Glycidyl-methylether
Formel: C₄H₈O₂
MDL: MFCD00273372
CAS-Nummer: 64491-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR