(S)-(+)-Glycidyl-methylether, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

454699-1G
SIAL454699-1GEA 128 EUR
SIAL454699-1G
(S)-(+)-Glycidyl-methylether, Sigma-Aldrich®
(S)-(+)-Glycidyl-methylether
Formel: C₄H₈O₂
MDL: MFCD00273366
CAS-Nummer: 64491-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR