(-)-Diisopropyl-L-malat, Sigma-Aldrich®

Lieferant: Merck

454877-25ML
SIAL454877-25MLEA 161 EUR
SIAL454877-25ML
(-)-Diisopropyl-L-malat, Sigma-Aldrich®
(-)-Diisopropyl-L-malat
Formel: C₁₀H₁₈O₅
Molecular Weight: 218,25 g/mol
MDL: MFCD00274300
CAS-Nummer: 83541-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR