(+)-Dimethyl-D-malat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D-(+)-Malic acid dimethyl ester, Dimethyl D-(+)-Malate, D-(+)-Apple acid dimethyl ester, (R)-Dimethyl 2-hydroxysuccinate

M-Clarity™ Qualitätsstufe = MQ100

455040-5ML
SIAL455040-5MLEA 272 EUR
SIAL455040-5ML
(+)-Dimethyl-D-malat, Sigma-Aldrich®
(+)-Dimethyl-D-malat
Formel: C₆H₁₀O₅
Molecular Weight: 162,1406 g/mol
MDL: MFCD00066216
CAS-Nummer: 70681-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR