1-Brom-4-(trimethylsilyl)benzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Bromophenyl)trimethylsilane

M-Clarity™ Qualitätsstufe = MQ100

455113-5G
SIAL455113-5GEA 167 EUR
SIAL455113-5G
1-Brom-4-(trimethylsilyl)benzol, Sigma-Aldrich®
1-Brom-4-(trimethylsilyl)benzol
Formel: C₉H₁₃BrSi
Molecular Weight: 229,1890 g/mol
MDL: MFCD00094165
CAS-Nummer: 6999-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR