Tricyclohexylphosphin 20% in Toluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

455164-100G
SIAL455164-100GEA 104 EUR
SIAL455164-100G
Tricyclohexylphosphin 20% in Toluol, Sigma-Aldrich®
Tricyclohexylphosphine
Formel: C₁₈H₃₃P
MDL: MFCD00003853
CAS-Nummer: 2622-14-2
EINECS: 220-069-2
UN: 1294
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR