(R)-(-)-1-Benzyl-3-hydroxypiperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-1-Benzyl-3-piperidinol

M-Clarity™ Qualitätsstufe = MQ100

455172-5G
SIAL455172-5GEA 229 EUR
SIAL455172-5G
(R)-(-)-1-Benzyl-3-hydroxypiperidin, Sigma-Aldrich®
(R)-(-)-1-Benzyl-3-hydroxypiperidin
Formel: C₁₂H₁₇NO
Molecular Weight: 191,27 g/mol
MDL: MFCD00274307
CAS-Nummer: 91599-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR