Hafniumtetrakis(dimethylazanid), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrakis(dimethylamino)hafnium, TDMAH

M-Clarity™ Qualitätsstufe = MQ100

455199-25G
SIAL455199-25GEA 1060 EUR
SIAL455199-25G
Hafniumtetrakis(dimethylazanid), Sigma-Aldrich®
Tetrakis(dimethylamino)hafnium(IV)
Formel: C₈H₂₄HfN₄
Molecular Weight: 354,793 g/mol
MDL: MFCD01862473
CAS-Nummer: 19782-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR