Hafniumtetrakis(diethylazanid), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

455202-10G
SIAL455202-10GEA 278 EUR
SIAL455202-10G
Hafniumtetrakis(diethylazanid), Sigma-Aldrich®
Tetrakis(diethylamino)hafnium(IV)
Formel: C₁₆H₄₀HfN₄
MDL: MFCD00799095
CAS-Nummer: 19824-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR