α,α,α,6-Tetrafluor-m-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluoro-5-(trifluoromethyl)benzoic acid

M-Clarity™ Qualitätsstufe = MQ100

455253-5G
SIAL455253-5GEA 43.7 EUR
SIAL455253-5G
α,α,α,6-Tetrafluor-m-toluylsäure, Sigma-Aldrich®
α,α,α,6-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
MDL: MFCD00061292
CAS-Nummer: 115029-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR