α,α,α,5-Tetrafluor-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

455334-1G
SIAL455334-1GEA 27 EUR
SIAL455334-1G
α,α,α,5-Tetrafluor-o-toluylsäure, Sigma-Aldrich®
α,α,α,5-Tetrafluor-o-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
MDL: MFCD00083521
CAS-Nummer: 654-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR