Hexachloroiridium(IV)säure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dihydrogen hexachloroiridate(IV) xhydrate (Chunks or pellets), Hydrogen iridium hexachloride, Hydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Dihydrogen hexachloroiridate(IV) hydrate
M-Clarity™ Qualitätsstufe = MQ100
455962-1G
SIAL455962-1GEA
271
EUR
SIAL455962-1G
Hexachloroiridium(IV)säure, Sigma-Aldrich®
Hexachloroiridic(IV) acid hydrate
Formel:
H₂Cl₆Ir·xH₂O Molecular Weight: 406,93 g/mol (anhy) |
MDL:
MFCD00149906 CAS-Nummer: 110802-84-1 EINECS: 241-012-8 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...