n-Nonadecan-[D40], Sigma-Aldrich®

Lieferant: Merck

Synonyme: n-Nonadecane-[²H₄₀ ]

M-Clarity™ Qualitätsstufe = MQ200

456276-250MG
SIAL456276-250MGEA 255 EUR
SIAL456276-250MG
n-Nonadecan-[D40], Sigma-Aldrich®
n-Nonadecan-[D40]
Formel: C₁₉D₄₀
Molecular Weight: 308,21 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 39756-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR