Fluoranthen-[D10] (98% D), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Fluoranthene-[²H₁₀]

M-Clarity™ Qualitätsstufe = MQ200

456292-100MG
SIAL456292-100MGEA 202 EUR
SIAL456292-100MG
Fluoranthen-[D10] (98% D), Sigma-Aldrich®
Fluoranthen-[D10]
Formel: C₁₆D₁₀
CAS-Nummer: 93951-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR