2,5-Dibrom-3-hexylthiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

456373-5G 456373-25G
SIAL456373-5GEA 97.9 EUR
SIAL456373-5G SIAL456373-25G
2,5-Dibrom-3-hexylthiophen, Sigma-Aldrich®
2,5-Dibromo-3-hexylthiophene
Formel: C₁₀H₁₄Br₂S
MDL: MFCD00274313
CAS-Nummer: 116971-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR