3-Cyclopenten-1-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: cyclopent-3-ene-1-carboxylic acid, Cyclopent-3-enecarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

456519-1G
SIAL456519-1GEA 30.1 EUR
SIAL456519-1G
3-Cyclopenten-1-carbonsäure, Sigma-Aldrich®
3-Cyclopenten-1-carbonsäure
Formel: C₆H₈O₂
Molecular Weight: 112,13 g/mol
MDL: MFCD00101764
CAS-Nummer: 7686-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR