4,4'-Biphenyldiboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: [1,1'-Biphenyl]-4,4'-diyldiboronic acid

M-Clarity™ Qualitätsstufe = MQ100

456799-500MG
SIAL456799-500MGEA 228 EUR
SIAL456799-500MG
4,4'-Biphenyldiboronsäure, Sigma-Aldrich®
4,4'-Biphenyldiboronsäure
Formel: C₁₂H₁₂B₂O₄
Molecular Weight: 241,85 g/mol
MDL: MFCD00151795
CAS-Nummer: 4151-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR