4-(Methylthio)phenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid
M-Clarity™ Qualitätsstufe = MQ100
456802-5G
SIAL456802-5GEA
66.6
EUR
SIAL456802-5G
4-(Methylthio)phenylboronsäure, Sigma-Aldrich®
4-(Methylthio)phenylboronsäure
Formel:
C₇H₉BO₂S Molecular Weight: 168.02 g/mol |
MDL:
MFCD00093410 CAS-Nummer: 98546-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...