2-Brom-4-(trifluormethoxy)anilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

457388-10G
SIAL457388-10GEA 20.4 EUR
SIAL457388-10G
2-Brom-4-(trifluormethoxy)anilin, Sigma-Aldrich®
2-Brom-4-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
MDL: MFCD00203478
CAS-Nummer: 175278-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR