N-Cbz-D-phenylalaninol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Benzyloxycarbonyl-D-phenylalaninol, Z-D-Phenylalaninol, Cbz-D-Phenylalaninol

M-Clarity™ Qualitätsstufe = MQ100

459933-5G
SIAL459933-5GEA 168 EUR
SIAL459933-5G
N-Cbz-D-phenylalaninol, Sigma-Aldrich®
N-Cbz-D-phenylalaninol
Formel: C₁₇H₁₉NO₃
Molecular Weight: 285,3 g/mol
MDL: MFCD00191193
CAS-Nummer: 58917-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR