1,2-Bis(diphenylphosphino)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

460273-1G
SIAL460273-1GEA 84.4 EUR
SIAL460273-1G
1,2-Bis(diphenylphosphino)benzol, Sigma-Aldrich®
1,2-Bis(diphenylphosphino)benzol
Formel: C₃₀H₂₄P₂
MDL: MFCD00014081
CAS-Nummer: 13991-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR