2-Iod-5-methylthiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

460532-5G
SIAL460532-5GEA 138 EUR
SIAL460532-5G
2-Iod-5-methylthiophen, Sigma-Aldrich®
2-Iod-5-methylthiophen
Formel: C₅H₅IS
Molecular Weight: 224,07 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00022492
CAS-Nummer: 16494-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR