1,2-Bis(carboxymethylthio)ethan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Ethylenedithio)diacetic acid, (Ethylenedithio)diaceticacid

46084-10G-F
SIAL46084-10G-FEA 82.3 EUR
SIAL46084-10G-F
1,2-Bis(carboxymethylthio)ethan, Sigma-Aldrich®
1,2-Bis(carboxymethylthio)ethan
Formel: C₆H₁₀O₄S₂
Molecular Weight: 210,26 g/mol
MDL: MFCD00021771
CAS-Nummer: 7244-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR