(R)-(+)-β-Methylphenethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(+)-beta-Methylphenethylamine, (R)-(+)-2-Phenyl-1-propylamine

M-Clarity™ Qualitätsstufe = MQ100

461385-1G
SIAL461385-1GEA 68.4 EUR
SIAL461385-1G
(R)-(+)-β-Methylphenethylamin, Sigma-Aldrich®
(R)-(+)-β-Methylphenethylamin
Formel: C₉H₁₃N
MDL: MFCD00216740
CAS-Nummer: 28163-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR