(R)-(+)-1-Phenyl-1-propanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(+)-(2-Hydroxy-1-methylethyl)benzol, (R)-(+)-β-Methyl-β-phenethylalkohol

M-Clarity™ Qualitätsstufe = MQ200

461407-1G
SIAL461407-1GEA 183 EUR
SIAL461407-1G
(R)-(+)-1-Phenyl-1-propanol, Sigma-Aldrich®
(R)-(+)-2-Phenyl-1-propanol
Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00145206
CAS-Nummer: 19141-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR