(S)-(-)-N-(1-Phenylethyl)succinamidsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-4-Oxo-4-((1-phenylethyl)amino)butanoic acid

M-Clarity™ Qualitätsstufe = MQ100

461431-5G
SIAL461431-5GEA 293 EUR
SIAL461431-5G
(S)-(-)-N-(1-Phenylethyl)succinamidsäure, Sigma-Aldrich®
(S)-(-)-N-(1-Phenylethyl)succinamidsäure
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221,26 g/mol
MDL: MFCD00077433
CAS-Nummer: 21752-34-1
EINECS: 244-569-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR