(R)-(+)-N-(1-Phenylethyl)phthalamidsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(+)-N-(1-Phenylethyl)phthalamic acid, (R)-(+)-N-(alpha-Methylbenzyl)phthalamic acid, (R)-2-((1-Phenylethyl)carbamoyl)benzoic acid

M-Clarity™ Qualitätsstufe = MQ100

461458-1G
SIAL461458-1GEA 71.8 EUR
SIAL461458-1G
(R)-(+)-N-(1-Phenylethyl)phthalamidsäure, Sigma-Aldrich®
(R)-(+)-N-(1-Phenylethyl)phthalamidsäure
Formel: C₁₆H₁₅NO₃
Molecular Weight: 269,3 g/mol
MDL: MFCD00063050
CAS-Nummer: 21752-35-2
EINECS: 244-570-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR