(S)-(-)-N-(1-Phenylethyl)phthalamidsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(-)-N-(alpha-Methylbenzyl)phthalamic acid, (S)-(-)-N-(1-Phenylethyl)phthalamic acid, (S)-2-((1-Phenylethyl)carbamoyl)benzoic acid

M-Clarity™ Qualitätsstufe = MQ100

461466-1G
SIAL461466-1GEA 67.7 EUR
SIAL461466-1G
(S)-(-)-N-(1-Phenylethyl)phthalamidsäure, Sigma-Aldrich®
(S)-(-)-N-(1-Phenylethyl)phthalamidsäure
Formel: C₁₆H₁₅NO₃
Molecular Weight: 269,3 g/mol
MDL: MFCD00013979
CAS-Nummer: 21752-36-3
EINECS: 244-571-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR