(1R)-1-Phenyl-N-[(1R)-1-phenylethyl]ethanaminhydrochlorid 97% (durch GC), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (R,R)-Bis(alpha-methylbenzyl)amine Hydrochloride, (R,R)-(+)-Bis(1-phenylethyl)amine Hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

461520-5G
SIAL461520-5GEA 168 EUR
SIAL461520-5G
(1R)-1-Phenyl-N-[(1R)-1-phenylethyl]ethanaminhydrochlorid 97% (durch GC), Sigma-Aldrich®
(R,R)-(+)-Bis(alpha-methylbenzyl)amine hydrochloride
Formel: C₁₆H₁₉N
Molecular Weight: 261,79 g/mol
Schmelzpunkt: 260…263 °C (Lit.)
MDL: MFCD00216672
CAS-Nummer: 82398-30-9

Bestellen

Spezifikation Testergebnisse

Infrared Spectrum Conforms to Structure
Optical Rotation (c = 3%, Ethanol) 71.0 - 75.0°
Titration by AgNO₃ 96.5 - 103.5%
Purity (GC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR