D(+)-Fenchon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(1S)-(+)-1,3,3-Trimethylbicyclo[2.2.1]-2-heptanon, (1S,4R)-(+)-1,3,3-Trimethylbicyclo[2.2.1]-2-heptanon
46210-100ML-F
SIAL46210-100ML-FEA
175
EUR
SIAL46210-100ML-F
D(+)-Fenchon, Sigma-Aldrich®
D(+)-Fenchon
Formel:
C₁₀H₁₆O Molecular Weight: 152,24 g/mol Siedepunkt: 203 °C (1013 hPa) Dichte: 0,9449 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070689 CAS-Nummer: 4695-62-9 EINECS: 225-160-0 UN: 1224 ADR: 3,III Merck Index: 13,03995 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...