(S)-(-)-2-Bromisovaleriansäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(-)-2-Bromo-3-methylbutyric acid

462101-5G
SIAL462101-5GEA 124 EUR
SIAL462101-5G
(S)-(-)-2-Bromisovaleriansäure, Sigma-Aldrich®
(S)-(-)-2-Bromisovaleriansäure
Formel: C₅H₉BrO₂
Molecular Weight: 181,03 g/mol
CAS-Nummer: 26782-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR