Methyl-4-piperidincarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-Piperidinecarboxylate, Methyl isonipecotate, Methyl piperidine-4-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

463485-25ML
SIAL463485-25MLEA 53.8 EUR
SIAL463485-25ML
Methyl-4-piperidincarboxylat, Sigma-Aldrich®
Methyl-4-piperidincarboxylat
Formel: C₇H₁₃NO₂
Molecular Weight: 143.18 g/mol
MDL: MFCD00190578
CAS-Nummer: 2971-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR