Dinatriumhexahydroxoplatinat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dinatriumplatinat, Natriumhexahydroxoplatinat(IV)
M-Clarity™ Qualitätsstufe = MQ100
464708-1G
SIAL464708-1GEA
147
EUR
SIAL464708-1G
Dinatriumhexahydroxoplatinat, Sigma-Aldrich®
Dinatriumhexahydroxoplatinat
Formel:
H₆Na₂O₆Pt Molecular Weight: 343,1 g/mol |
MDL:
MFCD00014236 CAS-Nummer: 12325-31-4 EINECS: 235-590-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...