(Cyanmethyl)triphenylphosphoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
464937-25G
SIAL464937-25GEA
111
EUR
SIAL464937-25G
(Cyanmethyl)triphenylphosphoniumchlorid, Sigma-Aldrich®
(Cyanomethyl)triphenylphosphonium chloride
Formel:
C₂₀H₁₇ClNP |
MDL:
MFCD00031672 CAS-Nummer: 4336-70-3 EINECS: 224-383-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...